Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

648744

Sigma-Aldrich

Mandelamide

97%

Synonym(s):

2-Hydroxy-2-phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH(OH)CONH2
CAS Number:
Molecular Weight:
151.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

133-137 °C (lit.)

SMILES string

NC(=O)C(O)c1ccccc1

InChI

1S/C8H9NO2/c9-8(11)7(10)6-4-2-1-3-5-6/h1-5,7,10H,(H2,9,11)

InChI key

MAGPZHKLEZXLNU-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[Molecular biology aspects of the antimicrobial effect of synthetic chemotherapeutic agents].
N Popov
Zeitschrift fur arztliche Fortbildung, 76(15), 662-666 (1982-08-01)
Clemens Lamberth et al.
Pest management science, 62(5), 446-451 (2006-03-22)
Novel types of anti-oomycetic compounds have been designed and prepared. The synthetic approach to these mandelamides is outlined. Biological data demonstrate their high efficacy against important plant diseases such as tomato and potato late blight (Phytophthora infestans De Bary) and
Zi Wei Luo et al.
Metabolic engineering, 62, 298-311 (2020-10-18)
Benzoic acid (BA) is an important platform aromatic compound in chemical industry and is widely used as food preservatives in its salt forms. Yet, current manufacture of BA is dependent on petrochemical processes under harsh conditions. Here we report the
Michael J McLeish et al.
Journal of bacteriology, 185(8), 2451-2456 (2003-04-03)
The enzymes of the mandelate metabolic pathway permit Pseudomonas putida ATCC 12633 to utilize either or both enantiomers of mandelate as the sole carbon source. The genes encoding the mandelate pathway were found to lie on a single 10.5-kb restriction
Biank T Gonçalves et al.
Magnetic resonance in chemistry : MRC, 46(5), 418-426 (2008-03-11)
Interesting anisotropic effects were observed for phenylglyoxamides and their respective mandelamides. Such effects were observed in experimental (1)H and (13)C NMR (in CDCl(3), CD(3)OD, and DMSO-d(6) solvents) and in some cases with good correlation to theoretical (1)H and (13)C NMR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service