MilliporeSigma
  • Anisotropic and hydrogen bonding effects in phenylglyoxamides and mandelamides: theoretical and NMR conformational evaluation.
Anisotropic and hydrogen bonding effects in phenylglyoxamides and mandelamides: theoretical and NMR conformational evaluation.
Magnetic resonance in chemistry : MRC (2008-03-11)
Biank T Gonçalves, Pierre M Esteves, Angelo C Pinto, Carlos R Kaiser, Fernanda L da Silva, Eduardo Miguez, Joaquim F M da Silva
ABSTRACT
Interesting anisotropic effects were observed for phenylglyoxamides and their respective mandelamides. Such effects were observed in experimental (1)H and (13)C NMR (in CDCl(3), CD(3)OD, and DMSO-d(6) solvents) and in some cases with good correlation to theoretical (1)H and (13)C NMR DFT-GIAO (B3LYP/6-311++G**//B3LYP/6-31G*) calculations. A systematic conformational analysis of these compounds was performed in a two-step methodology, using PM3 and DFT (B3LYP/6-31G*) calculations; with good accomplishment and computational time economy. It was observed that intramolecular hydrogen bonding plays a significant role in the conformation of such compounds. Finally, a geminal nonequivalence of an N-CH(2) moiety, in one of the alkyl side chain (R1 = R2), was found for the tertiary mandelamides studied.