Merck
Septa, Barrier, pkg 100 aluminum/silicone, diam. × thickness 11 mm × 0.060 in., for use with 4 mL vial
Septa, Barrier, pkg 100
Septa, Barrier, pkg 100 aluminum/silicone, diam. × thickness 11 mm × 0.060 in., for use with 4 mL vial