Documentos Principais
G005079
GC Analysis of (R)-(+)-1-Phenylethylamine (N-Trifluoroacetyl Derivative) on Astec® CHIRALDEX G-PN
suitable for GC
About This Item
técnica(s)
gas chromatography (GC): suitable
parâmetros de teste
column: Astec CHIRALDEX G-PN, 30 m x 0.25 mm I.D., 0.12 μm (74033AST)
oven: 130 °C
inj. temp.: 250 °C
detector: FID, 250 °C
carrier gas: helium, 30 psi
injection: 1 μL, 80:1 split
sample: 3 mg/mL in methanol
peak 1: (R)-(+)-1-phenylethylamine (N-TFA derivative) (ee% 99.1)
peak 2: (S)-(-)-1-phenylethylamine (N-TFA derivative)
aplicação(ões)
chemicals and industrial polymers
Informações legais
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC
coluna analítica
Escolha uma das versões mais recentes:
Certificados de análise (COA)