Documentos Principais
21912
SGE Syringes for Agilent/HP 7673, 7683 and 6850 ALS Autosampler, Fixed Needle
volume 10 μL, SK-10F-HP-0.47, needle size 26 ga, needle L 42 mm, pkg of 6 ea
About This Item
Código UNSPSC:
42142600
Materiais
cone tip stainless steel needle (point style 1)
tamanho da agulha
26 ga
Características
needle type F (fixed needle)
embalagem
pkg of 6 ea
fabricante/nome comercial
SGE
agulha C
42 mm
volume
10 μL
compatibilidade
for use with Agilent/HP 7673 and 7683 autosampler
Descrição geral
SGE syringes are designed and tested to meet critical autosampler specifications. These syringes are designed with a unique plunger to eliminate bending and seizing. The needle point style withstands multiple and fast septum injections, making it one of the most troublefree autosampler syringes available.
Escolha uma das versões mais recentes:
Certificados de análise (COA)