Documentos Principais
Z742662
Tube-O-Tank for use with the Tube-O-DIALYZER
small
About This Item
Código UNSPSC:
41123100
NACRES:
NB.24
embalagem
pkg of 1 ea
fabricante/nome comercial
G-BioSciences 786-145D
Descrição geral
Dialysis tank specifically designed for use with the Tube-O-DIALYZER. Two sizes are available that are suitable for Micro (Small) and Medi (Large) size Tube-O-DIALYZER Each tank will accommodate 7 Tube-O-DIALYZER and are supplied with floats.
Informações legais
Tube-O-DIALYZER is a trademark of Geno Technology, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)