Z692700
microDOC Compact gel documentation system, with UV Transilluminator (UVT254)
AC/DC input 230 V AC
About This Item
Código UNSPSC:
41105300
NACRES:
NB.22
Produtos recomendados
Características
Contains UK and Schuko plugs
fabricante/nome comercial
Cleaver CSI-MDOCUV254
Descrição geral
This option combines the microDOC Compact Gel Documentation System with a 12 x 12 cm UV 254 nm tranilluminator.
Available with optional TotalLab™ 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
12 x 12 cm UV 254 nm transilluminator
Available with optional TotalLab™ 1D software. Main software functions include: lane creation, background subtraction, band detection, profile deconvolution, and molecular size, pI and quantity calibration.
System comprises of:
Canon G12 digital
2MB storage card
Ethidium bromide & close up +3 filters
Lift-off hood with easy access door and built-in inner lights
8 in. TFT color monitor
12 x 12 cm UV 254 nm transilluminator
Informações legais
TotalLab is a trademark of Nonlinear Dynamics Ltd.
Escolha uma das versões mais recentes:
Certificados de análise (COA)