Documentos Principais
Z325015
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
one mark, capacity 9 mL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Materiais
AR-Glas®
descrição
total delivery Ex + 15 sec waiting time
Características
class AS
embalagem
pack of 6 ea
fabricante/nome comercial
BRAND 29711
capacidade
9 mL
Descrição geral
Additional to DIN
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Outras notas
BR29711 (India)
Informações legais
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG