OH30253070
Ohaus® Scout® Portable Balance
Model SKX123, weighing capacity 120 g, resolution: 0.001 g, AC/DC input 230 V, universal plug set
About This Item
Código UNSPSC:
41111502
NACRES:
NB.71
Produtos recomendados
Materiais
metal weighing plate
Características
CE compliant
embalagem
pack of 1 ea
fabricante/nome comercial
Ohaus® 30253070
capacidade de pesagem faixa de medição
120 g, resolution: 0.001 g
diâmetro do tacho de pesagem
9.3 cm
Descrição geral
Ideal for laboratory and industrial applications, the OHAUS Scout comes in a slim, stackable design with large backlit LCD. Features include superior overload protection, multiple weighing units and application modes. Geared for high performance in your facility with fast stabilization time and high resolution weighing results, these portable balances set a new standard in laboratory and industrial weighing.
Aplicação
Weighing, Parts Counting, Percent Weighing, Check Weighing, Totalization, Display Hold
Características e benefícios
Stability indicator, stackable design with optional storage cover (excluding 1mg models), mechanical and software overload/underload protection, low battery indicator, auto shut-off, auto tare, span calibration mass included on models up to 620g capacity
Informações legais
Ohaus is a registered trademark of Ohaus Corp.
Scout is a registered trademark of Ohaus Corp.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
OH30253070-1EA | 4065266747188 |