Documentos Principais
HS20613
Manual pipette racks
red plastic, double
About This Item
Código UNSPSC:
41122801
NACRES:
NB.35
Materiais
acrylonitrile-butadiene-styrene (ABS)
red plastic
Características
Holds 0.1 to 50 mL pipettes or cans
fabricante/nome comercial
Heathrow Scientific HS20613
L × P × A
291 mm × 88 mm × 401 mm
tamanho
291x88x401 mm
Descrição geral
Angled four shelved compartments keep your 0.1 to 50 mL pipettes or cans within easy reach, while preventing damage and clutter.