About This Item
Código UNSPSC:
41121800
NACRES:
NB.16
Produtos recomendados
Materiais
assorted colors
embalagem
pack of 100 ea
fabricante/nome comercial
Heathrow Scientific 120653
Descrição geral
The Slide Folder Clasps provide an area for identification labels and secure the slide folder so that your slides do not fall out when the folder is transported.
Características e benefícios
- Made from ABS
- Ideal for histology, microbiology and pathology as well as multiple user research environments
- Clasps can be labeled with a permanent marker or a label can be attached
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
HS120653-100EA | 4065265819619 |