EPM1289-0400
Test Tube Rack, for Eppendorf® New Brunswick™ Innova® and Eppendorf® Excella® Shakers
holds 30 tubes (18-21 mm diam.)
About This Item
Código UNSPSC:
41122804
NACRES:
NB.73
Produtos recomendados
Características
holds 30 tubes (18-21 mm diam.)
fabricante/nome comercial
Eppendorf® M1289-0400 (Global)
tamanho
450x80x129 mm
Categorias relacionadas
Descrição geral
Test Tube Racks have an adjustable angle and tube height support. Made from stainless-steel with inserts to help retain tubes and prevent them from spinning in the rack. Finger-tight knobs let you quickly adjust angle, or release the rack from the base plate, for easy transport to the bench.
Informações legais
Eppendorf is a registered trademark of Eppendorf AG
Excella is a registered trademark of Eppendorf, Inc.
Innova is a registered trademark of Eppendorf, Inc.
New Brunswick is a trademark of Eppendorf AG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
EPM1289-0400-1EA | 4061835081714 |