Key Documents
EP5820719001
Adapter for Eppendorf® FA-45-6-30 Rotor
capacity, 1 x 10 mL Oak Ridge tubes, pack of 2
About This Item
Código UNSPSC:
41103911
NACRES:
NB.82
Características
capacity 1 x 10 mL Oak Ridge tubes
embalagem
pack of 2
fabricante/nome comercial
Eppendorf® 5820719001 (Global)
técnica(s)
centrifugation: suitable
compatibilidade
for use with Rotor FA-45-6-30
Descrição geral
Adapters for various size tubes and bottles for use with Eppendorf rotor FA-45-6-30.
Informações legais
Eppendorf is a registered trademark of Eppendorf AG