Key Documents
EP5810756006
Eppendorf® Adapter for 250 mL Rectangular Bucket
for use with 8 x 7-18 mL tubes, set of 2 pieces
About This Item
Código UNSPSC:
41103911
NACRES:
NB.82
Características
for use with 8 x 7-18 mL tubes
embalagem
set of 2 pieces
fabricante/nome comercial
Eppendorf® 5810756006
técnica(s)
centrifugation: suitable
compatibilidade
for use with 250 mL rectangular buckets
Informações legais
Eppendorf is a registered trademark of Eppendorf AG