Key Documents
EP5810743001
Eppendorf® Swing-bucket rotor A-4-81
without buckets
About This Item
Código UNSPSC:
41103909
NACRES:
NB.82
Características
to hold 4 x 4 MTP/flex buckets
to hold 4 x 500 mL rectangular buckets
without buckets
embalagem
pack of 1 ea
fabricante/nome comercial
Eppendorf® 5810743001
Parâmetros
4,000 max. RCF (3,220 xg)
4,000 rpm (3,220 xg)
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models 5804/5804R/5810/5810R
Informações legais
Eppendorf is a registered trademark of Eppendorf AG