EP5810711002
Eppendorf® Swing-bucket rotor A-4-62-MTP
A-4-62-MTP with 4 MTP buckets
About This Item
Código UNSPSC:
41103909
NACRES:
NB.82
Características
to hold 4 x MTP buckets
A-4-62-MTP with 4 MTP buckets
embalagem
pack of 1 ea
fabricante/nome comercial
5810711002
Eppendorf® 5810711002
Parâmetros
4,000 max. RCF (3,220 xg)
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models 5804/5804R/5810/5810R
Descrição geral
- Eppendorf® Swing-bucket rotor accommodates tubes and bottles from 0.2 mL to 250 mL as well as plates.
- Aerosol-tight caps for safe centrifugation of hazardous samples optionally available.
- Rotor life time: max 40,000 cycles.
- Rotor, buckets, caps and adapters are autoclavable.
Informações legais
Eppendorf is a registered trademark of Eppendorf AG
Escolha uma das versões mais recentes:
Certificados de análise (COA)