Documentos Principais
EP5804750004
Eppendorf® Adapter For 100 mL Rectangular Bucket
holds standard, Vacutainer, Falcon and micro test tubes 1.2-5 mL, pack of 2 ea
About This Item
Código UNSPSC:
41103911
NACRES:
NB.82
Características
holds standard, Vacutainer, Falcon and micro test tubes 1.2-5 mL
embalagem
pack of 2 ea
fabricante/nome comercial
Eppendorf® 5804750004
técnica(s)
centrifugation: suitable
compatibilidade
for use with 100 mL retangular bucket
Descrição geral
Tube adapters for use with Eppendorf 100 mL rectangular bucket.
Informações legais
Eppendorf is a registered trademark of Eppendorf AG