Key Documents
EP5702740004
Eppendorf® Swing-bucket rotor A-4-38
without buckets
About This Item
Código UNSPSC:
41103909
NACRES:
NB.82
Características
without buckets (without buckets)
without buckets
embalagem
pack of 1 ea
fabricante/nome comercial
Eppendorf® 5702740004
Parâmetros
4,400 max. RCF (2,750 xg)
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models 5702/5702R/5702RH
Informações legais
Eppendorf is a registered trademark of Eppendorf AG