Key Documents
EP5427750002
Rotor for Eppendorf® 5430/5430R Centrifuges
FA-45-16-17, aerosol-tight, to hold, 16 x 5.0 mL Eppendorf tubes, with lid and wind shield
About This Item
Código UNSPSC:
41103909
NACRES:
NB.82
Características
to hold 16 x 5.0 mL Eppendorf tubes
aerosol-tight
fixed angle rotor
with lid and wind shield
embalagem
pack of 1 ea
fabricante/nome comercial
Eppendorf® 5427750002 (Global)
Parâmetros
14,200 rpm
21,190 × g max. RCF
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models 5430/5430R
Informações legais
Eppendorf is a registered trademark of Eppendorf AG