Key Documents
EP5424700004
Eppendorf® Rotor FA-45-24-11-Special
incl. lid, aerosol tight, coated
About This Item
Código UNSPSC:
41103909
NACRES:
NB.82
Características
to hold 24 x 1.5/2.0 ml tubes
incl. lid, aerosol tight, coated
embalagem
pack of 1 ea
fabricante/nome comercial
Eppendorf® 5424700004
Parâmetros
15,000 max. RCF (21,130 xg)
técnica(s)
centrifugation: suitable
compatibilidade
for use with centrifuge models 5424/5424R
Informações legais
Eppendorf is a registered trademark of Eppendorf AG