Documentos Principais
Z743177
Julabo® bath covers
lift-up gable cover to fit CORIO C-B13/17 and CD-B13/17
About This Item
Código UNSPSC:
41103706
NACRES:
NB.77
embalagem
pack of 1 ea
fabricante/nome comercial
Julabo 9970253
Descrição geral
The hinged stainless steel gable hood reduces the evaporation of tempering liquid and saves energy. The rising steam is led back into the bath via a drip edge. This means that the tempering liquid remains in the bath and refill cycles are extended. An additional benefit is that the hood prevents contamination of the samples in the bath. The hinged stainless steel gable hood is designed for operation up to +60°C.
Informações legais
Julabo is a registered trademark of Julabo USA, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)