Z742795
Kern EMB series precision balances
Kern EMB 200-3, weighing capacity 200 g, accuracy: 0.001 g, precision: ±0.005 g
Sinônimo(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
About This Item
Produtos recomendados
embalagem
pack of 1 ea
fabricante/nome comercial
Kern EMB 200-3
capacidade de pesagem faixa de medição
200 g, accuracy: 0.001 g, precision: ±0.005 g
Categorias relacionadas
Descrição geral
- Simple and convenient 2-key operation, making them ideal for use in schools and universities
- Tare function facilitates formulation work
- Particularly flat design
- Secure and non-slip positioning with rubber feet
- Ready for use: Batteries included. AUTO-OFF function to preserve the batteries, can be switched off
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z742795-1EA | 4061838807472 |