Z742418
Q-Tube™ Pressure Monitor
35 SS, pack of 1 ea
About This Item
Código UNSPSC:
41121500
NACRES:
NB.51
Produtos recomendados
Materiais
316 stainless steel adapter
PTFE sleeve
embalagem
pack of 1 ea
fabricante/nome comercial
Q Labtech 100-PRMSET-35SS
Parâmetros
180 psi max. pressure
Descrição geral
The parts with direct chemical contact are made of 316 stainless steel, PTFE, Simriz, or glass.
Pressure Monitor Set-35-SS/100-PRMSET-N35SS contains all items needed for immediate use (316 stainless steel Adapter and matched PTFE sleeve, 1 pressure gauge, 1 pressure tube-35 mL, 10 Septa-35 mL-180 psi, etc.).
Pressure Monitor Set-35-SS/100-PRMSET-N35SS contains all items needed for immediate use (316 stainless steel Adapter and matched PTFE sleeve, 1 pressure gauge, 1 pressure tube-35 mL, 10 Septa-35 mL-180 psi, etc.).
Informações legais
Q-Tube is a trademark of Q Labtech LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z742418-1EA | 4061834419624 |