Z682276
Ace rotary evaporator condenser complete "CR" assembly
glass, fits Büchi® models R-200/R-205 and R-114/R-144
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
Produtos recomendados
Materiais
glass
fabricante/nome comercial
Büchi equivalent, 11511
Descrição geral
This "CR" condenser assembly is similar to the "C" assembly except that the bottom joint is 45/40 inner to fit into a separate distribution head. The complete "CR" assembly consists of the main glass outer trap body, glass cold trap inner body, PTFE sealing ring with Viton® O-rings, PTFE lid and hose barbs with GL threaded caps.
Informações legais
Büchi is a registered trademark of Büchi Labortechnik AG
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Escolha uma das versões mais recentes:
Certificados de análise (COA)