Z677574
Lapren™ latex/chloroprene flock lined gloves
size XL
About This Item
Código UNSPSC:
46181504
NACRES:
NB.35
Materiais
green (chloroprene, natural latex)
tamanho
XL
Características
CE compliant
disposable
standard type EN 374 (cat. III)
standard type EN 388 (cat. III)
embalagem
pack of 100 ea
fabricante/nome comercial
KCL 706 - 10
KCL 706
C × espessura
310 mm × 0.6 mm
Descrição geral
Lapren incorporates a high quantity of polychloroprene into latex to improve chemical resistance. A mixture of raw materials ensures a high elasticity as well as good tensile strength and tear resistance. The palm grip surface ensures a firm grip even in wet working environments without loss of the high degree of sensitivity.
With cotton flock lining, stretched cuff and anatomical design, this product meets all requirements of comfort and hygiene.
With cotton flock lining, stretched cuff and anatomical design, this product meets all requirements of comfort and hygiene.
- Ergonomic fit and high elasticity ensures good sensitivity
- Profiled palm for firm grip when wet
- High mechanical resistance
- Cotton flock lining improves comfort
- Free from allergenic substances such as thiurames, benzothiazoles and thiourea
- CE compliant
Informações legais
Lapren is a trademark of KCL GmbH
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z677574-100EA | 4061833069820 |