Z675997
RF1000™ pipette filler
a lightweight manual pipette filler uniquely designed for one-handed operation
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Produtos recomendados
Materiais
plastic body
silicone bulb
fabricante/nome comercial
Heathrow Scientific HS1000RFU
tamanho
180 mm × 45 mm × 120 mm
volume
1-100 mL
Categorias relacionadas
Descrição geral
Heathrow Scientifics RF1000 is a lightweight manual pipette filler uniquely designed for one-handed operation. Simply squeeze the main bulb, and push the toggle forward to aspirate or back to dispense. Push the blowout bulb to expel any liquid remaining in the pipette.
The RF1000 accommodates graduated or volumetric pipettes (glass or plastic) and can be used with volumes ranging from 1 mL to 100 mL. Includes a hydrophobic 0.45 μm filter and a magnetic holder for metal surface mount or wall mount. The polypropylene nozzle is autoclavable.
The RF1000 accommodates graduated or volumetric pipettes (glass or plastic) and can be used with volumes ranging from 1 mL to 100 mL. Includes a hydrophobic 0.45 μm filter and a magnetic holder for metal surface mount or wall mount. The polypropylene nozzle is autoclavable.
Informações legais
RF1000 is a trademark of Heathrow Scientific
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z675997-1EA | 4061832769639 |