Z566772
Scienceware® Secador® desiccator cabinet
model 3.0, vertical profile, amber
About This Item
Código UNSPSC:
41104510
NACRES:
NB.43
Produtos recomendados
Materiais
polycarbonate
Características
Can be stacked three high.
embalagem
pack of 1 ea
fabricante/nome comercial
model 3.0
Bel-Art F42073-1008
A × L × P
41.7 cm × 34.1 cm × 41.4 cm
abertura da porta A × L
31.2 cm × 22.5 cm
cor
amber
Descrição geral
Dry storage of humidity sensitive products is now easier. Secador cabinets assure moisture and dust free storage of valuable items like reagents and pipettes with their proprietary sealed design.
Available in small or large models, with either horizontal or vertical orientation, these uniquely-designed units are available totally clear, clear with blue end caps, and amber. The stackable small model is perfect for maximizing storage space in a minimum footprint.
Cabinets are molded of polycarbonate resistant to staining, crazing and chemical attack and have built-in protection that blocks 99% of UV light. Desiccants can be placed under the bottom shelf or on any shelf. A hygrometer in the door allows for easy monitoring of relative humidity. Carrying handles make them easy to relocate.
Amber cabinets reduce visible light penetration by more than 50% and blocks 99% of UV light.
Available in small or large models, with either horizontal or vertical orientation, these uniquely-designed units are available totally clear, clear with blue end caps, and amber. The stackable small model is perfect for maximizing storage space in a minimum footprint.
Cabinets are molded of polycarbonate resistant to staining, crazing and chemical attack and have built-in protection that blocks 99% of UV light. Desiccants can be placed under the bottom shelf or on any shelf. A hygrometer in the door allows for easy monitoring of relative humidity. Carrying handles make them easy to relocate.
Amber cabinets reduce visible light penetration by more than 50% and blocks 99% of UV light.
- Has two perforated shelves that are removable
- Door has two latches at the top and loops for - lock or tamper evident seal
Outras notas
Model 1.0 has 1 fixed shelf and 1 door latch. Models 3.0 and 4.0 have 3 removable shelves.
Informações legais
Scienceware is a registered trademark of SP Scienceware, a division of SP
Secador is a registered trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z566772-1EA | 4061832265919 |