Z556378
Aldrich® Sure/Stor™ adapter
joint: ST/NS 24/40
About This Item
Código UNSPSC:
41121815
NACRES:
NB.43
Produtos recomendados
Junta
joint: ST/NS24/40
descrição
with PTFE J Young valve and SafetyBarb®
hose connector
Categorias relacionadas
Descrição geral
Converts round-bottom flasks to Sure/Stor flasks.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Sure/Stor is a trademark of Sigma-Aldrich Co. LLC
parte de substituição
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z556378-1EA | 4061832113166 |