Documentos Principais
Z556351
Sigma-Aldrich
Aldrich® Sure/Stor adapter
joint: ST/NS 19/22
About This Item
Código UNSPSC:
41121815
NACRES:
NB.43
Junta
joint: ST/NS19/22
descrição
with PTFE J Young valve and SafetyBarb®
hose connector
Descrição geral
Converts round-bottom flasks to Sure/Stor flasks.
NOTICE: We supply high quality PTFE valves on Aldrich brand glassware that has been tested and approved by our Glass Center.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
SafetyBarb is a registered trademark of Merck KGaA, Darmstadt, Germany
Sure/Stor is a trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)