Z543209
Periodic table of the elements
Poster size
About This Item
Código UNSPSC:
60104506
NACRES:
CE.34
Produtos recomendados
L × C
24 in. × 36 in.
Descrição geral
Laminated. Excellent reference tools for students and researchers.
Poster:
Full-color. Chart information includes: atomic weight, melting point, boiling point, density, electronegativity, first ionization potential, group classifications, atomic number, oxidation states, element symbol, electronic configuration, and name. Plus special tables for Elementary Subatomic Particles and % Ionic Character of a Single Chemical Bond.
Binder:
Two-sided chart features more than 20 properties of the elements, from electron configuration and electronegativity to covalent radius and thermal conductivity.
Wallet:
Two-sided chart features properties of the elements in table format with useful chemical constants on the back.
Poster:
Full-color. Chart information includes: atomic weight, melting point, boiling point, density, electronegativity, first ionization potential, group classifications, atomic number, oxidation states, element symbol, electronic configuration, and name. Plus special tables for Elementary Subatomic Particles and % Ionic Character of a Single Chemical Bond.
Binder:
Two-sided chart features more than 20 properties of the elements, from electron configuration and electronegativity to covalent radius and thermal conductivity.
Wallet:
Two-sided chart features properties of the elements in table format with useful chemical constants on the back.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z543209-1EA | 4061833454862 |