Z415626
Aldrich® dual bank manifold with high-vacuum PTFE valves
manifold with vacuum-gauge port, positions, 3
About This Item
Código UNSPSC:
40161501
NACRES:
NB.79
Produtos recomendados
Características
positions 3
Comprimento
300 mm
Categorias relacionadas
Descrição geral
The dual bank manifolds come with 0-10 mm bore high-vacuum PTFE valves.
Attention: We supply high quality PTFE valves on Aldrich brand glassware; these have been tested and approved by our Glass Center.
Attention: We supply high quality PTFE valves on Aldrich brand glassware; these have been tested and approved by our Glass Center.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
produto relacionado
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z415626-1EA | 4061832111407 |