Z266094
TLC plate storage racks
Rack for 20 cm × 20 cm plates
About This Item
Código UNSPSC:
41115711
NACRES:
NC.07
Produtos recomendados
Materiais
stainless steel (20 gauge)
técnica(s)
thin layer chromatography (TLC): suitable
Categorias relacionadas
Descrição geral
TLC plate storage racks is a rack of convenient size, which is used to hold the prepared plates during the process of drying and transportation. It may be used to store the agar plates used in agarose gel electrophoresis.
Used to carry and store TLC plates without damage to media. Racked plates may be air or oven dried. Holds up to 10 standard plates in either a vertical or horizontal position. Racks are stainless steel with a carrying handle.
produto relacionado
Nº do produto
Descrição
Preços
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Laboratory diagnosis of specific organ diseases by means of combined serum isoenzyme patterns.
Nerenberg TS and Pogojeff G
American Journal of Clinical Pathology, 51(4), 429-439 (1969)
Food Chemicals Codex (2003)
Global Trade Item Number
SKU | GTIN |
---|---|
Z266094-1EA | 4061833634127 |