Z260274
FORTUNA® Universal Optifix® HF dispenser
HF model, volume 6-30 mL
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Produtos recomendados
Características
subdivision 1.0 mL
fabricante/nome comercial
Fortuna Graf 101.106-44
faixa de medição de volume
6-30 mL
volume
6-30 mL
conector
GL 40
GL 45
Categorias relacionadas
Descrição geral
For all liquids that can be handled with the Optifix dispensers and for hydrofluoric acid (HF)
Features:
Supplied:
FORTUNA Universal Optifix HF dispenser, with 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Features:
- Dispensing cylinder made of PTFE
- Massive PTFE encased dispensing piston
- FIX adapter made of PTFE
- Ergonomic finger protection cap
- Fast and easy volume setting with additional fine adjustment
- Valve block made of PTFE, with additional safety valve in the ventilation tube, no metal springs in the valves - no corrosion
Supplied:
FORTUNA Universal Optifix HF dispenser, with 2 PTFE adapters GL40 + GL45 & 1 PTFE intake tube
Informações legais
FORTUNA is a registered trademark of Poulten & Graf Ltd.
Optifix is a registered trademark of Poulten & Graf Ltd.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
Z260274-1EA | 4061833568989 |