Z259551
Ace photochemical immersion well
Quartz
About This Item
Código UNSPSC:
41121800
NACRES:
NC.25
Produtos recomendados
fabricante/nome comercial
Ace Glass 787435
Categorias relacionadas
Descrição geral
Double-walled, with small diameter inner tube that extends down the annular space to insure flow of coolant from bottom of well upwards to outlet.
Compression seal inside reactor vessel allows adjustable height of well. Total length: 390 mm.
Compression seal inside reactor vessel allows adjustable height of well. Total length: 390 mm.
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
Z259551-1EA | 4061826748053 |