Z174831
ACE microscale glassware kit
MICRO/MINI-Lab® Deluxe kit
About This Item
Código UNSPSC:
41100000
NACRES:
NB.43
Produtos recomendados
fabricante/nome comercial
Ace Glass 9560-14
Categorias relacionadas
Descrição geral
Designed to give students experience with a wide range of organic reactions with increased safety. Kits contain microscale glassware needed to perform experiments described in the textbook Microscale Organic Laboratory with Multistep and Multiscale Syntheses.
Greaseless, threaded ST14/10 connections can be interfaced with traditional ST14/10 joints. Conical reaction vials include open-top caps, septa, and O-rings. Glassware in the DELUXE kit is supplied in a foam-lined plastic carrying case. The BASIC kit is supplied in a clear plastic, compartmentalized box with tilt lid.
Greaseless, threaded ST14/10 connections can be interfaced with traditional ST14/10 joints. Conical reaction vials include open-top caps, septa, and O-rings. Glassware in the DELUXE kit is supplied in a foam-lined plastic carrying case. The BASIC kit is supplied in a clear plastic, compartmentalized box with tilt lid.
Informações legais
Micro/Mini-Lab is a registered trademark of Ace Glass, Inc.
produto relacionado
Nº do produto
Descrição
Preços
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Xi Chen et al.
Immunobiology, 221(1), 48-55 (2015-08-27)
Macrophages function as an essential component of innate immune system, contributing to both the initiation and appropriate resolution of inflammation. The exposure of macrophages to the microbial products, such as lipopolysaccharide (LPS), can strongly shift the balance between tissue homeostasis
Xia Pan et al.
Food chemistry, 188, 569-575 (2015-06-05)
Polygalacturonase is one of the most important industrial pectinases. To enrich the genetic resources and develop new enzyme candidates, three polygalacturonase genes (Nfpg I-III) of glycosyl hydrolase family 28 were cloned from Neosartorya fischeri P1 and functionally expressed in Pichia
Ze-Quan Mo et al.
Fish & shellfish immunology, 47(1), 280-288 (2015-09-20)
CCR6 have been demonstrated playing an important role in immune cells homing to mucosal tissues, mediating antigen presentation and immune response in mammals. CCR6 in lower vertebrate leukocyte homing has not yet been revealed. Cryptocaryon irritans is believed to be
Global Trade Item Number
SKU | GTIN |
---|---|
Z174831-20X1KT | |
Z174831-1EA | |
Z174831-1KT | 4061832348070 |