Documentos Principais
Z171387
Supelco
Series A-2 gas syringe
capacity 25 μL
About This Item
Código UNSPSC:
41122001
NACRES:
NB.03
Materiais
bevel tip stainless steel needle
tamanho da agulha
2 in.
Características
needle bevel tip
fabricante/nome comercial
VICI 050023
capacidade
25 μL
agulha C
2 in.
Descrição geral
Unique Sample-Lok push-button valve allows sample storage in syringes as small as 25μL and permits pressurization of samples prior to injection, for better resolution. Plunger, when fully depressed, makes flush contact with bottom of syringe barrel, virtually eliminating dead volume. Parts are replaceable.
Escolha uma das versões mais recentes:
Certificados de análise (COA)