Z168130
Ampule breaker/collar
For 11-20 mL ampules
About This Item
Código UNSPSC:
41121807
NACRES:
NB.43
Descrição geral
How to Use:
1. Slip polyethylene breaker/ collar over top of ampule.
2. Grip bottom of ampule with one hand and breaker/collar with other.
3. Gently snap top of ampule off.
4. Discard collar and ampule top.
1. Slip polyethylene breaker/ collar over top of ampule.
2. Grip bottom of ampule with one hand and breaker/collar with other.
3. Gently snap top of ampule off.
4. Discard collar and ampule top.
Escolha uma das versões mais recentes:
Certificados de análise (COA)