Z167517
Aldrich® Schlenk-type Vigreux distilling column
male joint: ST/NS 14/20, female joint: ST/NS 14/20, L 150 mm
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
Produtos recomendados
rótulo
O
Junta
female joint: ST/NS14/20
male joint: ST/NS14/20
Comprimento
150 mm
Categorias relacionadas
Descrição geral
Replacement part for Aldrich Air-Sensitive Glass kit. Thermoplastic screw caps with Viton O-rings usable to 140 deg C. Vigreux indents run the entire length of the column and act as the packing for the column.
Ligação
See the Threaded Ground Glass Tutorial to see how threaded joints work.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Artigos
Versatile system for chemical reactions, extractions, and transfers of solids and liquids in laboratories.
Global Trade Item Number
SKU | GTIN |
---|---|
Z167517-1EA | 4061837609442 |