Documentos Principais
Z158062
Sigma-Aldrich
Poly-jaque lab jack
polypropylene, plate size 6 in. × 6 in.
About This Item
Código UNSPSC:
41122800
NACRES:
NB.35
Materiais
polypropylene
fabricante/nome comercial
Bel-Art
tamanho da placa
6 in. × 6 in.
Descrição geral
Nonsparking and nonmagnetic, corrosion-resistant. Self-lubricating with polyether-imide/PTFE bushings. Working height to 11in. Maximum static load is 15lb.
Informações legais
Poly-jaque is a trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Visite a Biblioteca de Documentos
Os clientes também visualizaram
Slide 1 of 7
1 of 7
Swiss Boy lab jacks load ≤50 kg (middle position), platform L × W 120 mm × 140 mm, Model 110
Z635332
Swiss Boy lab jacks
Support jack platform L × W 30 cm × 30 cm
Z565857
Support jack
Support jack platform L × W 24 cm × 24 cm
Z565849
Support jack
Swiss Boy lab jacks load ≤80 kg (with two plates, middle position), platform L × W 200 mm × 200 mm, Model 115
Z635545
Swiss Boy lab jacks
Swiss Boy lab jacks load ≤30 kg (middle position), platform L × W 80 mm × 75 mm, Model 105
Z635324
Swiss Boy lab jacks
Swiss Boy lab jacks load ≤90 kg (middle position), platform L × W 180 mm × 216 mm, Model 116
Z635359
Swiss Boy lab jacks
Swiss Boy lab jacks load ≤90 kg (middle position), platform L × W 240 mm × 320 mm, Model 118
Z635383
Swiss Boy lab jacks
Global Trade Item Number
SKUGTIN
Z158062-1EA4061832256948
BAF182040000-1EA4065269235552