Z126616
Orbital-attachment kit
for building models in three dimensions
About This Item
Código UNSPSC:
41100000
NACRES:
NB.17
Produtos recomendados
Descrição geral
Lightweight orbitals for building models in three dimensions. Parts are finished in standard colors and attach to the atom centers contained in the giant-size molecular model kits. Pi bonds fit over ends of orbitals to show overlap.
Kit contains a selection of 50 parts, and is useful for studying elementary and advanced concepts of orbital theory. Storage box has individual compartments for each orbital.
Kit contains a selection of 50 parts, and is useful for studying elementary and advanced concepts of orbital theory. Storage box has individual compartments for each orbital.
Escolha uma das versões mais recentes:
Certificados de análise (COA)