PL800GA-NIT-9
Plas-Labs replacement nitrile gloves
for 2 piece glove set, size 9 (for 8 in. ports)
About This Item
Código UNSPSC:
46181504
NACRES:
NB.35
Produtos recomendados
Materiais
nitrile
tamanho
9 (for 8 in. ports)
Características
reusable
for use with glove box sleeves
embalagem
pack of 1 pair
fabricante/nome comercial
Plas-Labs 800GA-NIT-9
Descrição geral
Plas-Labs glovebox gloves are available as either one-piece or two-piece assemblies with interchangeable sleeves and gloves. This enables personnel to use the same glove box by changing the size or type of glove being used. Choose from four rubber types. Gloves are 15 mil thickness unless noted otherwise.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
Note: Order sleeves, gloves, and glove over sleeve connectors separately.
- White Hypalon® rubber for resistance to abrasion, oxidizing chemicals, ozone and UV light
- Black neoprene rubber for chemical resistance
- Black butyl rubber for highest impermeability to water vapor, gases, and toxic fluids
- Green nitrile rubber for resistance to oil, fuel, and acids
- Latex rubber for flexibility
Informações legais
Hypalon is a registered trademark of E. I. du Pont de Nemours and Company
Escolha uma das versões mais recentes:
Certificados de análise (COA)