Documentos Principais
PL800CARTGB
Plas-Labs transport support cart
fits GB, NB, or AC style glove boxes
About This Item
Código UNSPSC:
41103406
NACRES:
NB.35
Materiais
stainless steel
Características
Anti-scuff casters including brakes and locks
embalagem
pkg of 1 ea
L × P × A
51 in. × 24 in. × 35 in.
Descrição geral
Suspended sliding shelf for single operator units. Culture plate capacity is about 340. Fabricated of .375 in. thick clear acrylic, with shelves spaced at 7 in. apart. The entire assembly is suspended from the chamber top to keep the floor area uncluttered.
All carts have anti-scuff casters including brakes and locks. Carts also feature a cushioning system that minimizes vibration, shocks, and wear and tear on chambers.
Escolha uma das versões mais recentes:
Certificados de análise (COA)