ESYNTH017
SynLectro™ Aluminum Electrode
size 3 mm × 20 mm × 60 mm
About This Item
Código UNSPSC:
81141502
NACRES:
NA.06
Produtos recomendados
tamanho
3 mm × 20 mm × 60 mm
Categorias relacionadas
Descrição geral
SynLectro™ Aluminum Electrode is a 3x20x60 mm electrode designed for use with the SynLectro™ Electrolysis Platform for synthetic electrochemistry.
SynLectro™ Electrolysis Platform
The SynLectro™ Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
SynLectro™ Electrolysis Platform
The SynLectro™ Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component, designed with consistency, reproducibility, and safety in mind.
Informações legais
SynLectro is a trademark of Merck KGaA, Darmstadt, Germany
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
ESYNTH017-1EA | 4065265641258 |