Documentos Principais
DWK73750-13100
KIMBLE® disposable screw thread culture tube with white marking spot
round bottom borosilicate glass, tube diam. × L (13 mm) (100 mm), non-sterile
About This Item
Código UNSPSC:
41121706
NACRES:
NB.11
Materiais
round bottom borosilicate glass
round bottom
screw top neck
transparent borosilicate glass tube
esterilidade
non-sterile
Características
without cap
embalagem
case of 1000 ea Modular tray shrink-wrapped in plastic film
fabricante/nome comercial
Kimble® 73750-13100
diâmetro do tubo × C
(13 mm) (100 mm)
diâmetro do tubo
(13 mm)
conector
neck (External Thread)
Descrição geral
Disposable Screw Thread Culture Tubes with Marking Spot
  • These culture tubes are suitable for tissue culture work and general bacteriological use.
  • Round-bottomed
  • Each tube has a chevron marking area
  • Supplied without caps
  • Manufactured from 51 expansion borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class B requirements
Informações legais
KIMBLE is a registered trademark of DWK Life Sciences
Escolha uma das versões mais recentes:
Certificados de análise (COA)