CAD6524
1-way threaded end adapter (UTS)
MLL to 1/4-28 standard thread (316SS)
About This Item
Código UNSPSC:
41121815
NACRES:
NB.03
Produtos recomendados
embalagem
pack of 5 ea
fabricante/nome comercial
Cadence Science 6524
Categorias relacionadas
Descrição geral
- Plated brass or 316 stainless steel
- Meets ISO 594 specifications
- Final assembly water tested at 45 PSI
Female Luer Lock (FLL) - contains a tabbed hub for secure fitting to a threaded male Luer lock.
Micro-Mate® Female Luer Lock (MMFLL) - designed and manufactured to reduce the stripping of threads which occurs when metal adapters are mated with plastic male Luer lock adapters. Uniquely designed to facilitate easy cleaning and maintenance.
Male Luer Lock (MLL) - male Luer slip with a threaded sheath for a secure fitting to a female Luer lock.
Male Luer (ML) - male Luer slip that fits with a female Luer.
Informações legais
Micro-Mate is a registered trademark of Cadence Science
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
CAD6524-5EA | 4061832517919 |