BR723015
BRAND® Haemacytometer cover glass, for counting chambers
borosilicate glass, size 20 mm × 26 mm × 0.4 mm
Sinônimo(s):
Cover glasses, Microscopy
About This Item
Código UNSPSC:
41122600
NACRES:
NB.22
Produtos recomendados
Materiais
borosilicate glass
Características
IVD
fabricante/nome comercial
BRAND 723015
tamanho
20 mm × 26 mm × 0.4 mm
tipo de ligação
non-treated surface
Categorias relacionadas
Descrição geral
Borosilicate glass, DIN ISO 8255. Refractive index ne = 1.52 ± 0.01, Abbe number ve = 56.5 ± 0.5. Flatness tolerance ± 3 μm.
They differ from ordinary cover glasses by their plain ground and polished surface.
They differ from ordinary cover glasses by their plain ground and polished surface.
Embalagem
2 cover glasses in a plastic bag, separated by tissue paper.
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
BR723015-100EA | 4061831859003 |