BR705930
BRAND® Transferpette® S Multichannel Pipette
capacity (20-200 μL), 12-channel, adjustable volume, M12-200, DE-M
Sinônimo(s):
Pipettes, Piston-operated pipettes, Positive displacement principle, Repair set, Transferpettor
About This Item
Produtos recomendados
Características
12-channel adjustable volume
embalagem
case of 1 ea
fabricante/nome comercial
BRAND 705930
capacidade
(20-200 μL)
Categorias relacionadas
Descrição geral
Overview- Volume range of single-channel pipette: from 0.1 μl to 10 ml- Volume range of multi-channel pipette: from 0.5 μl to 300 μl- Short stroke and low operating forces- Universal tip cone- Completely autoclaving at 121 μC- One-hand operation for right- and left-handed users- Color code on volume adjustment wheel, shaft and volume-change protection- Easy insertion and ejection of tips due to O-rings on multi-channel pipettes- Can be calibrated easily and without tools thanks to Easy Calibration technology- Slim shaft for sampling from narrow vessels (centrifuge tubes)- Operating temperature from +15 μC to +40 μC- Vapor pressure up to 500 mbar- Viscosity up to 260 mPa s
Efficient: One-hand operation
Flexible: ergonomic design for all hand sizes
Reliable: volume-change protection prevents accidental adjustments
Informações legais
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR705930-1EA | 4061841857365 |