BR704850
BRAND® TRANSFERPETTE® S starter kits
About This Item
Código UNSPSC:
41121500
NACRES:
NB.01
Produtos recomendados
fabricante/nome comercial
BRAND
Descrição geral
The Transferpette S microliter pipettes are manual single and multi-channel pipettes for productive and precise pipetting in the laboratory. They offer high quality and many features that make work easier and more efficient, for example: Genuine one-hand operation, short stroke, low operating forces, volume adjustment protection and an easy-to-read volume display. They are easy to maintain and can be adjusted without tools using Easy Calibration technology. The single-channel Transferpette S pipettes cover the volume range from 0.1 ul to 10 ml. The Transferpette S -8 and -12 multichannel pipettes are available in volumes from 0.5 ul to 300 ul.
Características e benefícios
- Efficient: one-hand operation
- Flexible: ergonomic design for all hand sizes
- Reliable: volume-change protection prevents accidental adjustments
Informações legais
BRAND is a registered trademark of BRAND GMBH + CO KG
Transferpette is a registered trademark of BRAND GMBH + CO KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Global Trade Item Number
SKU | GTIN |
---|---|
BR704850-5EA |