BAF199170150
Scienceware® vacuum aspirator collection system
capacity 3200 mL, AC/DC input 115 V AC
Sinônimo(s):
bottle aspirator system
About This Item
Código UNSPSC:
41100000
Produtos recomendados
embalagem
pack of 1 ea
fabricante/nome comercial
Bel-Art F19917-0150
capacidade
3200 mL
Categorias relacionadas
Descrição geral
Perform vacuum aspiration of a variety of liquids, including tissue culture media, supernatants, and chemical wastes. To use, simply insert a pipette tip, Pasteur pipette, or serological pipette into the collection tube and plug in to a 110/120V outlet. Self-contained system with bottle, tubing and vacuum pump is lightweight, compact and easy to use.
Características e benefícios
- Built-in pump quickly aspirates fluids while maintaining a low noise level (76db at 1 meter) and minimal vibration
- Ideal for fluid collection from tissue and/or cell culture, Petri dishes, plates, flasks and tubes
- Chemical and corrosion resistant polypropylene collection bottle holds 3800 mL (1 gallon) fluid wastes and has a safety release vent to prevent vacuum implosion
- A replaceable hydrophobic filter seals itself if it becomes wet protecting the pump motor from fluid contamination
- Compact design will not crowd workspace whether working in a hood or at the bench
Informações legais
Scienceware is a registered trademark of SP Scienceware, a division of SP
parte de substituição
Nº do produto
Descrição
Preços
Escolha uma das versões mais recentes:
Certificados de análise (COA)
Os clientes também visualizaram
Global Trade Item Number
SKU | GTIN |
---|---|
BAF199170150-1EA | 4061832872162 |