BAF182040000
SP Bel-Art® Single Burette Clamp
for Poly-Jaque™ Plastic Lab Jack Z158062
About This Item
Código UNSPSC:
41122800
NACRES:
NB.51
Produtos recomendados
Características
to hold Single
embalagem
pack of 1 ea
fabricante/nome comercial
Bel-Art F18204-0000
Descrição geral
Non-corroding clamps hold burettes firmly in place.
Características e benefícios
- Soft rubber pads hold burette without scratching
- Clamp fits standard 12.5mm (½") support rod and includes an adapter to fit to narrower 9.5mm (⅜") rod (support rod not included)
- Made of chemical resistant polypropylene; autoclavable at 121°C (250°F)
Informações legais
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Poly-jaque is a trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)